
3-(Methoxycarbonyl)phenylboronic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99769-19-4
:
Formula:C₈H₉BO₄
MDL Number:MFCD02093046
:179.97 g/mol
Melting Pt:187…191 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- HPLC:≥96 %
Specification Test Results
Frequently Bought Together