Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) ≥99%, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:623-33-6
:
Formula:C₄H₁₀ClNO₂
MDL Number:MFCD00012871
:139.58 g/mol
:M-Clarity™ quality level = MQ200
EINECS:210-787-4
Frequently Bought Together