
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:623-33-6
:
Formula:C₄H₁₀ClNO₂
MDL Number:MFCD00012871
:139.58 g/mol
Melting Pt:144…148 °C
EINECS:210-787-4
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together