Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dimethylamine
Dimethylamine 2 mol/l in methanol, Sigma-Aldrich®
Dimethylamine 2 mol/l in methanol, Sigma-Aldrich®
SIAL388246-100ML
:
Merck
:
Dimethylamine 2 mol/l in methanol, Sigma-Aldrich®
SIAL388246-100ML
:
Merck
:
Pack type:
Glass bottle
Pk:
100 mL
Seq:
2440
:
:
45.08 g/mol
:
Ambient
:
204-697-4
:
1992
:
3,II
:
124-40-3
:
C₂H₇N
:
MFCD00008288