Order Entry
Czech Republic
ContactUsLinkComponent
Herbicide reference standard, Propanil (3',4'-Dichloropropionanilide) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Herbicide reference standard, Propanil (3',4'-Dichloropropionanilide) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL16410000AL
Supplier:  EHRENSTORFER
CAS Number:  
Herbicide reference standard, Propanil (3',4'-Dichloropropionanilide) 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL16410000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    709-98-8
  • Description:
    Propanil (3',4'-Dichloropropionanilide) 10 µg/ml in acetonitrile
  • Formula:
    C₉H₉Cl₂NO
  • Description:
    Propani

Specifications

About this item