Herbicide reference standard, Monolinuron (N-(4-Chlorophenyl)-N'-methoxy-N'-methylurea) 500 µg/ml in methanol
Katalogové číslo PROOSL20207
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:1746-81-2
- Description:Monolinuron (N-(4-Chlorophenyl)-N'-methoxy-N'-methylurea) 500 µg/ml in methanol
- Formula:C₉H₁₁ClN₂O₂
- Description:Monolinuron