Cyanamide 50% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:420-04-2
:
Formula:CH₂N₂
MDL Number:MFCD00007572
:42.04 g/mol
:M-Clarity™ quality level = MQ100
EINECS:206-992-3
Frequently Bought Together