VWR®, Malathion, pesticide reference standard (1,2-Bis(ethoxycarbonyl)ethyl O,O-dimethyl phosphorodithioate), 100 µg/ml in methanol
Catalog # 124312W
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:1 mL
- CAS No.:121-75-5
- Description:Malathion (1,2-Bis(ethoxycarbonyl)ethyl O,O-dimethyl phosphorodithioate) 100 µg/ml in methanol
- Formula:C₁₀H₁₉O₆PS₂
- Description:Malathion (1,2-Bis(ethoxycarbonyl)ethyl O,O-dimethyl phosphorodithioate)