

Hexachloroiridic(IV) acid hexahydrate (∼40% Ir)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16941-92-7
:
Formula:H₂Cl₆Ir·6H₂O
MDL Number:MFCD00149907
Storage Temperature:Ambient
EINECS:241-012-8
UN:3260
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Appearance:Black Powder or crystals
- Assay:37 to 43 % (Ir)
Specification Test Results
Frequently Bought Together