

Manganese(II) perchlorate hexahydrate, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15364-94-0
:
Formula:Cl₂MnO₈·6H₂O
MDL Number:MFCD00150255
:361.931 g/mol
Melting Pt:165 °C (dec.)
Storage Temperature:Ambient
EINECS:237-390-9
UN:1481
ADR:5.1,II
:GHS03
:GHS07
Related Information
Frequently Bought Together