
2-Imidazolidinone hemihydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:121325-67-5
:
Formula:C₃H₆N₂O·0.5H₂O
MDL Number:MFCD02662355
:95.10 g/mol (86.09 anhy)
Melting Pt:55 °C
EINECS:204-436-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together