


Ethylmagnesium bromide ∼3.2 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:925-90-6
:
Formula:C₂H₅BrMg
MDL Number:MFCD00000043
:133.27 g/mol
Density:1.14 g/cm³
Flash Pt:–11 °C
EINECS:213-127-3
UN:3399
ADR:4.3,I
:GHS05
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark brown Solution (May develop some turbidity or precipitate)
- Titration Grignard:39 to 41 %
- Water:≤1 %
- Solubility:(2 % in DMSO) Clear colorless solution
- HPLC:≥97.5 %
Specification Test Results
Related Information
Frequently Bought Together