Organic reference standard, Fluorene 200 µg/ml in isooctane, CIL
Catalog # PROOCILULM74141.2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:86-73-7
- Description:Fluorene 200 µg/ml in isooctane
- Formula:C₁₃H₁₀
- Description:Fluorene