Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standardy
Standardy organické
2,4'-DDT 100 µg/ml
2,4'-DDT 100 µg/ml
Catalog # PROOCILULM61341.2
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
2,4'-DDT 100 µg/ml
Catalog # PROOCILULM61341.2
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Typ balení:
Glass bottle
Bal.:
1,2 mL
Čí. CAS:
789-02-6
Popis:
2,4'-DDT 100 µg/ml in n-nonane
Vzorec:
C₁₄H₉Cl₅
Popis:
2,4'-DDT
Specifications
About this item
Product Details
Product Details