Pesticide reference standard, γ-BHC 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM61331.2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:58-89-9
- Description:γ-BHC 100 µg/ml in n-nonane
- Formula:C₆H₆Cl₆
- Description:γ-BHC