
Diethyl octylphosphonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1068-07-1
:
Formula:C₁₂H₂₇O₃P
MDL Number:MFCD00015659
:250.32 g/mol
Boiling Pt:88…89 °C (0.1 mmHg)
Density:0.953
EINECS:213-941-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together