Diethyl decylphosphonate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16165-68-7
:
Formula:C₁₄H₃₁O₃P
MDL Number:MFCD00015593
Boiling Pt:128…130 °C (0.2 mmHg)
:278.38 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together