
3-(3,4-Difluorophenyl)propionic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:161712-75-0
:
Formula:C₉H₈F₂O₂
MDL Number:MFCD00799520
:186.16 g/mol
Melting Pt:49…53 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together