

3-(3,5-Dibromophenyl)propionic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:923977-15-5
:
Formula:C₉H₈Br₂O₂
MDL Number:MFCD11973898
Melting Pt:62 °C
:307.97 g/mol
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together