

Copper(II) formate tetrahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5893-61-8
:
Formula:C₂H₂CuO₄·4H₂O
MDL Number:MFCD00167128
Merck Index:14,2639
ADR:9,III
:225.65 g/mol (153.59 anhy)
EINECS:208-865-8
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together