Order Entry
Czech Republic
ContactUsLinkComponent
 
Flammable
Irritant
Environment
DL-Limonene for synthesis, Sigma-Aldrich®
Catalog # 8.14546.1000
Supplier:  Merck
CAS Number:  
Flammable
Irritant
Environment
DL-Limonene for synthesis, Sigma-Aldrich®
Catalog # 8.14546.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    -89 °C
  • Molar Mass:
    136.24 g/mol
  • Boiling Point:
    178 °C (1013 hPa)
  • Vapor Pressure:
    2.1 hPa (20 °C)

Specification Test Results

About this item

 :
 :136.24 g/mol
Merck Index:12,05518
Boiling Pt:176 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.8462 g/cm³ (20 °C)
Flash Pt:43 °C
Storage Temperature:Ambient
EINECS:205-341-0
UN:2052
ADR:3,III
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS07
 :GHS09
CAS Number:138-86-3
 :7705-14-8
Formula:C₁₀H₁₆
MDL Number:MFCD00062992