Order Entry
Czech Republic
ContactUsLinkComponent
Nitric acid 65% for trace analysis
 
 
Nitric acid 65% for trace analysis
  SCARAC16131000
 :  Scharlau
 :  
 
 
Nitric acid 65% for trace analysis
  SCARAC16131000
 :  Scharlau
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Pk:
    1.000 mL
  • Seq:
    2865

 

 

 :
 :63.01 g/mol
 :13,06608
 :120.5 °C (1013 hPa)
 :–42 °C
 :1.4 g/cm³ (20 °C)
 :Ambient
 :231-714-2
 :2031
 :8,II
 :GHS03
 :GHS05
 :7697-37-2
 :HNO₃
 :MFCD00011349