Mercury standard solution, 1000 mg/L Hg in 10% nitric acid standard for ICP
Katalogové číslo REGEPHG2A6
Supplier: REAGECON
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:10% HNO₃
- Pack type:Glass bottle
- Pk:100 mL
- Element:Mercury
- Symbol:Hg