Sleeves for ground joints, with gripping ring, BOLA
Supplier: Bohlender
Certificates
About this item
Reusable sleevesmade of PTFE, with gripping ring and sealing rings on the outside.
- Chemical resistant and temperature resistant from −200 to +250°C
- High-vacuum tight sealing
- Low leak rate: <0,0001 mbar× L/s
- Solid construction and sealing rings are easy accessible
Frequently Bought Together