Cheese trier
Supplier: GERBER FUNKE
Certificates
About this item
Chrome-nickel steel. Developed for sampling small cores and testing the readiness of cheeses without cutting them open. Ideal for use in milk-processing industries and cheese dairies.
- Black plastic handle
- Two different sizes available
Frequently Bought Together