
Citric acid monohydrate ≥98% for decalcifying
84559.290
:
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Assay:Min. 98 %
:
:210.14 g/mol
:13,02350
:135…152 °C
:1.552 g/cm³ (20 °C)
:173.9 °C (closed cup)
:201-069-1
:GHS07
:Ambient
:5949-29-1
:C₆H₈O₇.H₂O
:MFCD00149972