Order Entry
Czech Republic
ContactUsLinkComponent
Cutter for plastic tubing, SP Bel-Art
Cutter for plastic tubing, SP Bel-Art
Catalog # 229-0013
undefined
Cutter for plastic tubing, SP Bel-Art
Catalog # 229-0013

Specifications

  • Description:
    Cutter for flexible plastic tubing, 76 mm long
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

Portable, pocket-sized cutter cuts flexible tubing from 1,6 to 19 mm Ø.

  • Spring-loaded jaw will remain closed for safety when not in use
  • Replaceable, hardened steel blade makes clean and straight cuts