Magnetic sink strainer, Spinbar®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Spinbar® Magnetic Sink Strainer is a simple solution to prevent your Spinbar® Magnetic Stirring Bars from going down the sink drain.
- Four strong magnets attract and hold stir bars
- Four legs on the underside prevent strainer from being dislodged by flowing liquid
- Long slits in the sides and skirt allow fluids to drain rapidly
- Wide skirt allows use in a variety of sink drains
- Straight edge on one side allows it to be placed near the sink wall
- Flexible and chemical-resistant
Available in blue or green.
Frequently Bought Together