Eswabs collection kit
Supplier: Copan
Certificates
About this item
Universal open platform for bacterial culture and the preservation of DNA, RNA and antigens of diverse microorganisms. Features unique molded breakpoint with coloured indication mark and swab capture cap.
- Sample rapidly elutes into liquid phase
- Provides 1 ml for multitest analysis
- Maintains viability of aerobic, anaerobic and fastidious bacteria
Preserves nucleic acids and antigens of bacteria, viruses and Chlamydia.
Certifications: Certified DNase and RNase free.
Delivery: 50 kits per shelf pack – 500 kits per box.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together