Order Entry
Czech Republic
ContactUsLinkComponent
 
Desiccator cabinets, small stacking, dry-keeper™, SP Bel-Art
Catalog # 468-4101
undefined
Desiccator cabinets, small stacking, dry-keeper™, SP Bel-Art
Catalog # 468-4101

Specifications

  • W×D×H ext.:
    225×210×170 mm
  • Description:
    Small stacking desiccator, 4 l
  • Pk:
    1
  • With:
    1 chamber

Specifications

About this item

Small, clear polystyrene desiccator cabinet, these can be safely stacked up to five units high for maximum use of dry storage space.

Delivery: Supplied with ready to use jar of silica gel.