Filtering tiles, BOLA
Supplier: Bohlender
Jump to:
About this item
These filtering tiles are made of microporous PTFE.
- Suitable for cutting or stamping
- Resistant to chemicals
- Temperature resistance from –200 to +250 °C
4 Options Available Below
Product Details & Documents
These filtering tiles are made of microporous PTFE.
- Suitable for cutting or stamping
- Resistant to chemicals
- Temperature resistance from –200 to +250 °C
Product Family Options
Product Information
- MaterialDimensionsPore sizeThicknessAvailabilityPrice