Copper standard solution, concentrate, 1000 mg Cu in water (from CuCl₂) Titrisol® standard for AAS, Supelco®
Catalog # 1.09987.0001
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic ampoule
- Pk:1 Ampoul
- Seq:01
- M-Clarity quality level:MQ100
- Element:Copper
- Symbol:Cu
Specifications
About this item
Certifications: Traceable to SRM from NIST