(S)-(-)-1,1'-Bi-2-naphthol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:18531-99-2
:
Formula:C₂₀H₁₄O₂
MDL Number:MFCD00004068
:286.33 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ100
UN:2811
Frequently Bought Together