Order Entry
Czech Republic
ContactUsLinkComponent
(±)-1,2,3,4-Tetrahydro-2-naphthol 97%
Irritant
(±)-1,2,3,4-Tetrahydro-2-naphthol 97%
Certificates
undefined
Irritant
(±)-1,2,3,4-Tetrahydro-2-naphthol 97%

Specification Test Results

  • Appearance:
    Clear light yellow to light brown Viscous liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5630 to 1.5670 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together