Order Entry
Czech Republic
ContactUsLinkComponent
Thulium standard solution, 1000 mg/L Tm in 5% nitric acid (from Tm₂O₃), standard for AAS, Specpure®
Thulium standard solution, 1000 mg/L Tm in 5% nitric acid (from Tm₂O₃), standard for AAS, Specpure®
Catalog # 89889.AE
CAS Number:  
Thulium standard solution, 1000 mg/L Tm in 5% nitric acid (from Tm₂O₃), standard for AAS, Specpure®
Catalog # 89889.AE
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00011281
  • Element:
    Thulium
  • Symbol:
    Tm

Specifications

About this item

Molecular Formula: Tm₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011281

Certifications: Traceable to NIST