

Chloromethyl methyl sulphide 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2373-51-5
:
Formula:C₂H₅ClS
MDL Number:MFCD00000923
Boiling Pt:104…105 °C
Density:1.160
Flash Pt:17 °C (62 °F)
EINECS:219-148-4
ADR:3,II
:96.58 g/mol
:GHS02
:GHS07
Related Information
Frequently Bought Together