Order Entry
Czech Republic
ContactUsLinkComponent
 
Dichlofenthion(O-2,4-dichlorophenyl-O,O-diethyl phosphorothioate), Supelco®
Catalog # 45432-250MG
Supplier:  Merck
CAS Number:  
undefined
Dichlofenthion(O-2,4-dichlorophenyl-O,O-diethyl phosphorothioate), Supelco®
Catalog # 45432-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :315.16 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:202-564-5
CAS Number:97-17-6
Formula:C₁₀H₁₃Cl₂O₃PS