Order Entry
Czech Republic
ContactUsLinkComponent
 
Dichlofenthion (O-2,4-Dichlorophenyl O,O-diethyl phosphorothioate)
Catalog # EHERC12290000
Supplier:  EHRENSTORFER
CAS Number:  
Dichlofenthion (O-2,4-Dichlorophenyl O,O-diethyl phosphorothioate)
Catalog # EHERC12290000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :315.16 g/mol
EINECS:202-564-5
CAS Number:97-17-6
Formula:C₁₀H₁₃Cl₂O₃PS