(2-Hydroxyethyl)hydrazine, Sigma-Aldrich®
Catalog # SIAL54340-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:76.1 g/mol
EINECS:203-711-6
UN:2810
ADR:6.1,II
CAS Number:109-84-2
Formula:C₂H₈N₂O
MDL Number:MFCD00007623