
Sodium hexachlororhodate (III) dodecahydrate (17.1% Rh)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14972-70-4
:
Formula:Cl₆Na₃Rh·12H₂O
MDL Number:MFCD00003509
:600.77 g/mol
Melting Pt:900 °C (dec.)
EINECS:239-047-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together