Rhenium standard solution, 10000 mg/L Re in 5% nitric acid (from Re), standard for ICP, Specpure®
Molecular Formula: Re in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011195
: Traceable to NIST
- MDL No.:MFCD00011195
Frequently Bought Together