
trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3588-17-8
:
Formula:C₆H₆O₄
MDL Number:MFCD00002702
:142.11 g/mol
Merck Index:14,6299
Melting Pt:∼ 300 °C (dec.)
EINECS:222-724-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together