
trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid) 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3588-17-8
:
Formula:C₆H₆O₄
MDL Number:MFCD00002702
:142.11 g/mol
Boiling Pt:320 °C (760 mmHg)
Melting Pt:290 °C (dec.)
Storage Temperature:Ambient
EINECS:222-724-8
:GHS07
Specification Test Results
- Appearance:Beige Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Titration with NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together