
trans,trans-2,4-Dodecadienal ≥90% remainder mainly trans, cis isomer
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21662-16-8
:
Formula:C₁₂H₂₀O
MDL Number:MFCD00014678
:180.29 g/mol
Boiling Pt:130 °C (6 mmHg)
Density:0.863
Flash Pt:104 °C (219 °F)
EINECS:244-517-1
:GHS07
Related Information
Frequently Bought Together