
5,6,7,8-Tetrahydro-2-naphthol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1125-78-6
:
Formula:C₁₀H₁₂O
MDL Number:MFCD00001738
:148.2 g/mol
Boiling Pt:275…276 °C
Melting Pt:60…63 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:214-410-4
:GHS07
Related Information
Frequently Bought Together