Order Entry
Czech Republic
ContactUsLinkComponent
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) 95%
Irritant
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) 95%
Certificates
undefined
Irritant
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) 95%

Specification Test Results

  • Appearance:
    Clear colorless to yellow to green Liquid or low melting solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5550 to 1.5650 (20°C, 589 nm)
  • GC:
    ≥94 %

Specification Test Results

Related Information

Frequently Bought Together