Chromium standard solution, 1000 mg/L Cr in 5% hydrochloric acid (from Cr), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Cr in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00010944
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00010944
Specifications
Related Information
Frequently Bought Together