(2,6-Difluorophenyl)acetone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:101712-20-3
:
Formula:C₉H₈F₂O
MDL Number:MFCD02258867
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4790 to 1.4820 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together