Order Entry
Czech Republic
ContactUsLinkComponent
Zinc standard solution, 10 mg/L Zn in 5% nitric acid (from Zn), standard for ICP, Specpure®
Zinc standard solution, 10 mg/L Zn in 5% nitric acid (from Zn), standard for ICP, Specpure®
Catalog # 45269.AE
CAS Number:  
Zinc standard solution, 10 mg/L Zn in 5% nitric acid (from Zn), standard for ICP, Specpure®
Catalog # 45269.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • MDL No.:
    MFCD00011291
  • Element:
    Zinc
  • Symbol:
    Zn

Specifications

About this item

Molecular Formula: Zn in 2% HNO₃
Physical Form: Liquid
MDL No.: MFCD00011291

Certifications: Traceable to NIST