Order Entry
Czech Republic
ContactUsLinkComponent
Hydrogen fluoride pyridine complex (HF 65%) for synthesis, Sigma-Aldrich®
Toxic
Corrosive
Hydrogen fluoride pyridine complex (HF 65%) for synthesis, Sigma-Aldrich®
Catalog # 8.18068.0100
Supplier:  Merck
CAS Number:  
Toxic
Corrosive
Hydrogen fluoride pyridine complex (HF 65%) for synthesis, Sigma-Aldrich®
Catalog # 8.18068.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.14 g/cm3 (20 °C)
  • pH:
     - 7 (H2O, 20 °C)

Specification Test Results

About this item

 :
ADR:8 (6.1),I
 :M-Clarity™ quality level = MQ200
EINECS:231-634-8
UN:1790
 :GHS06
 :GHS05
Storage Temperature:Refrigerator
CAS Number:62778-11-4
Formula:C₅H₅N·(HF)x
MDL Number:MFCD00012436